accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNHHNNHOOOOH2CCH2HOOHCH3H3CCH3H3C(Z)(Z)

CAS RN 635-65-4

IUPAC Name: 3-(2-{[3-(2-carboxyethyl)-5-{[(2Z)-4-ethenyl-3-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-2-yl]methyl}-5-{[(2Z)-3-ethenyl-4-methyl-5-oxo-2,5-dihydro-1H-pyrrol-2-ylidene]methyl}-4-methyl-1H-pyrrol-3-yl)propanoic acid
Synonyms: bilirubin bilirubin ixα
Molecular Weight (g/mol): 584.67
Molecular Formula: C33H36N4O6
InChi Key: BPYKTIZUTYGOLE-IFADSCNNSA-N
SMILES: CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (4)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (7)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)