accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClO

CAS RN 635-93-8

IUPAC Name: 5-chloro-2-hydroxybenzaldehyde
Synonyms: 5-chloro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: FUGKCSRLAQKUHG-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1C=O
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)