CAS RN 63545-54-0
IUPAC Name:
3-[4-(chlorosulfonyl)phenyl]propanoic acid
Synonyms:
3-[4-(chlorosulfonyl)phenyl]propanoic acid
Molecular Weight (g/mol):
248.68
Molecular Formula:
C9H9ClO4S
InChi Key:
INJMPXHPNFJMLG-UHFFFAOYSA-N
SMILES:
OC(=O)CCC1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
3-(4-Chlorosulfonylphenyl)propionic acid, 99%
CAS: 63545-54-0 Molecular Formula: C9H9ClO4S Molecular Weight (g/mol): 248.68 MDL Number: MFCD02179510 InChI Key: INJMPXHPNFJMLG-UHFFFAOYSA-N Synonym: 3-4-chlorosulfonyl phenyl propanoic acid,benzenepropanoicacid, 4-chlorosulfonyl,3-4-chlorosulphonyl phenyl propanoic acid,3-4-chlorosulfonylphenyl propionic acid,3-4-chlorosulfonylphenyl propanoic acid,p-chlorosylphonyldihydrocinnamic acid,4-chlorosulfonyl benzenepropanoic acid,4-chlorosulphonyl dihydrocinnamic acid,p-chlorosulphonyldihydrocinnamic acid PubChem CID: 2757915 IUPAC Name: 3-[4-(chlorosulfonyl)phenyl]propanoic acid SMILES: OC(=O)CCC1=CC=C(C=C1)S(Cl)(=O)=O