accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrCH3NHH3CH3C

CAS RN 636-70-4

IUPAC Name: triethylazanium bromide
Synonyms: triethylamine, hydrobromide
Molecular Weight (g/mol): 182.11
Molecular Formula: C6H16BrN
InChi Key: NRTLTGGGUQIRRT-UHFFFAOYSA-N
SMILES: [Br-].CC[NH+](CC)CC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results