accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PPOCH3H3CH3C

CAS RN 638132-66-8

IUPAC Name: 1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethyl-1λ⁵-phospholan-1-one
Synonyms: 1-[2-(2,5-dimethylphospholan-1-yl)phenyl]-2,5-dimethyl-1λ⁵-phospholan-1-one
Molecular Weight (g/mol): 322.37
Molecular Formula: C18H28OP2
InChi Key: ORZVYNTUPREFTI-UHFFFAOYNA-N
SMILES: CC1CCC(C)P1C1=CC=CC=C1P1(=O)C(C)CCC1C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results