accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CMgCl

CAS RN 6393-56-2

IUPAC Name: magnesium(2+) pentan-1-ide chloride
Synonyms: magnesium(2+) pentan-1-ide chloride
Molecular Weight (g/mol): 130.9
Molecular Formula: C5H11ClMg
InChi Key: JFWWQYKSQVMLQU-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CCCC[CH2-]
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results