missing translation for 'onlineSavingsMsg'
Learn More

1-Pentylmagnesium chloride, 1M in MeTHF, Thermo Scientific™

Product Code. 15488067
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488067 500 mL 500mL
15478067 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488067 Supplier Thermo Scientific Alfa Aesar Supplier No. H51175.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6393-56-2
Molecular Formula C5H11ClMg
Molecular Weight (g/mol) 130.90
MDL Number MFCD00672089
InChI Key OHLJPYMGJFINNA-UHFFFAOYSA-M
Synonym pentylmagnesium chloride, pentylmagnesium chloride solution, n-pentylmagnesium chloride, 1-pentylmagnesium chloride, chloro pentyl magnesium, ohljpymgjfinna-uhfffaoysa-m, 1-pentylmagnesium chloride, 1m in methf, pentylmagnesium chloride solution, 2.0 m in thf
PubChem CID 10953547
IUPAC Name magnesium;pentane;chloride
SMILES CCCCC[Mg]Cl

Specifications

Linear Formula CH3(CH2)4MgCl
Quantity 500 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 130.9
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material 1-Pentylmagnesium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.