accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFON

CAS RN 63968-85-4

IUPAC Name: 2-(trifluoromethoxy)benzonitrile
Synonyms: 2-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 187.12
Molecular Formula: C8H4F3NO
InChi Key: ACNBBQGAWMHXLA-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC=C1C#N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results