CAS RN 64118-81-6
CAS RN 64118-81-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Diclofenac Acyl-Beta-D-glucuronide, TRC
CAS: 64118-81-6 Molecular Formula: C20 H19 Cl2 N O8 Molecular Weight (g/mol): 472.27 Synonym: β-D-Glucopyranuronic acid, 1-[2-[(2,6-dichlorophenyl)amino]benzeneacetate],Diclofenac glucuronide,Diclofenac-β-D-glucuronide IUPAC Name: (2S,3S,4S,5R,6S)-6-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid SMILES: O[C@@H]1[C@@H](O)[C@H](OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl)O[C@@H]([C@H]1O)C(=O)O