missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Acyl-Beta-D-glucuronide, TRC

Product Code. 30007867
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007867 10 mg 10mg
30004327 5 mg 5mg
30005318 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007867 Supplier Toronto Research Chemicals Supplier No. TRCD43647510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64118-81-6
Molecular Formula C20 H19 Cl2 N O8
Molecular Weight (g/mol) 472.27
Synonym β-D-Glucopyranuronic acid, 1-[2-[(2,6-dichlorophenyl)amino]benzeneacetate], Diclofenac glucuronide, Diclofenac-β-D-glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-6-[2-[2-(2,6-dichloroanilino)phenyl]acetyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@H](OC(=O)Cc2ccccc2Nc3c(Cl)cccc3Cl)O[C@@H]([C@H]1O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H19Cl2NO8/c21-10-5-3-6-11(22)14(10)23-12-7-2-1-4-9(12)8-13(24)30-20-17(27)15(25)16(26)18(31-20)19(28)29/h1-7,15-18,20,23,25-27H,8H2,(H,28,29)/t15-,16-,17+,18-,20+/m0/s1
Formula Weight 471.0488
Percent Purity >95%
Chemical Name or Material Diclofenac Acyl-beta-D-glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.