accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFBr

CAS RN 64248-56-2

IUPAC Name: 2-bromo-1,3-difluorobenzene
Synonyms: 2-bromo-1,3-difluorobenzene
Molecular Weight (g/mol): 192.99
Molecular Formula: C6H3BrF2
InChi Key: HRZTZLCMURHWFY-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1Br
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)