missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,6-difluorobenzene, 98%, ACROS Organics™

Product Code. 10655521 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10655521 5 g Glass bottle 5g
10241143 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10655521 Supplier Thermo Scientific Acros Supplier No. 297390050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 64248-56-2
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.991
MDL Number MFCD00009894
InChI Key HRZTZLCMURHWFY-UHFFFAOYSA-N
Synonym 1-bromo-2,6-difluorobenzene, 2,6-difluorobromobenzene, benzene, 2-bromo-1,3-difluoro, 2-bromo-1,3-difluoro-benzene, m-difluorobromobenzene, pubchem3034, acmc-1bc5o, 2,6-difluoro-bromobenzene, 1-bromo-2,6-diflurobenzene
PubChem CID 123557
IUPAC Name 2-bromo-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)Br)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2C6H3Br
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
Quantity 5 g
Formula Weight 192.99
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 6-difluorobenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.