accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 64275-73-6

H3COH(Z)

CAS RN 64275-73-6

IUPAC Name: (5Z)-oct-5-en-1-ol
Synonyms: (5Z)-oct-5-en-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: VDHRTASWKDTLER-ARJAWSKDSA-N
SMILES: CC\C=C/CCCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results