Learn More
cis-5-Octen-1-ol, 95%, remainder mainly trans-isomer, Thermo Scientific™
Product Code. 11464037
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11464037
Brand: Thermo Scientific Alfa Aesar A19454.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64275-73-6
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00015569
InChI Key VDHRTASWKDTLER-ONEGZZNKSA-N
Synonym oct-5-en-1-ol, e-oct-5-en-1-ol, 5-octen-1-ol, 5e-oct-5-en-1-ol
PubChem CID 6008958
IUPAC Name (E)-oct-5-en-1-ol
SMILES CCC=CCCCCO
Specifications
Density 0.85
Boiling Point 95°C (25 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.452
Quantity 25 g
Beilstein 1840670
Formula Weight 128.22
Percent Purity 95%
Assay remainder mainly trans-isomer
Chemical Name or Material cis-5-Octen-1-ol