CAS RN 64520-05-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
10-Hydroxy (E)-Amitriptyline, TRC
CAS: 64520-05-4 Molecular Formula: C20H23NO Molecular Weight (g/mol): 293.4 Synonym: (5E)-5-[3-(dimethylamino)propylidene]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol SMILES: CN(C)CC\C=C\1/c2ccccc2CC(O)c3ccccc13