accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3PCH3CH3CH3H3C

CAS RN 6476-36-4

IUPAC Name: tris(propan-2-yl)phosphane
Synonyms: triisopropylphosphine
Molecular Weight (g/mol): 160.24
Molecular Formula: C9H21P
InChi Key: IGNTWNVBGLNYDV-UHFFFAOYSA-N
SMILES: CC(C)P(C(C)C)C(C)C
Boiling Point 
  • (6)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (6)