missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylphosphine, 98%

Product Code. 11985841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11985841 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985841 Supplier Thermo Scientific Acros Supplier No. 317330250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6476-36-4 | C9H21P | 160.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6476-36-4
Density 0.8390g/mL
Boiling Point 81.0°C (22.0 mmHg)
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H21P
Linear Formula [(CH3)2CH]3P
Refractive Index 1.4690 to 1.4710
MDL Number MFCD00015029
Quantity 25 g
Synonym triisopropylphosphine, tri-i-propylphosphine, phosphine, triisopropyl, phosphine, tris 1-methylethyl, tri-isopropylphosphine, triisopropylphosphane, trisisopropylphosphine, triisopropyl phosphine, acmc-20ajdu, pubchem6458
InChI Key IGNTWNVBGLNYDV-UHFFFAOYSA-N
SMILES CC(C)P(C(C)C)C(C)C
IUPAC Name tri(propan-2-yl)phosphane
Molecular Weight (g/mol) 160.241
PubChem CID 80969
Formula Weight 160.24
Percent Purity 98%
Chemical Name or Material Triisopropylphosphine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.