accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHON

CAS RN 6480-68-8

IUPAC Name: quinoline-3-carboxylic acid
Synonyms: cinchoninic acid, 3-hydroxy-
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: DJXNJVFEFSWHLY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)