missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Quinoline-3-carboxylic acid, 98%

Product Code. 11318166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318166 5 g 5g
11308166 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318166 Supplier Alfa Aesar™ Supplier No. L15133.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6480-68-8
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00006770
InChI Key DJXNJVFEFSWHLY-UHFFFAOYSA-N
Synonym 3-quinolinecarboxylic acid, 3-quinoline carboxylic acid, 3-quinolinecarboxylicacid, pubchem7517, 3-quinolylcarboxylic acid, acmc-1bfc6, 3-quinolinecarboxylic acid 1g, 3-quinolinecarboxylic acid, purum hplc, q3c
PubChem CID 80971
IUPAC Name quinoline-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C=C(C=N2)C(=O)O

Specifications

Melting Point ∽283°C (decomposition)
Quantity 5 g
Beilstein 126542
Formula Weight 173.17
Percent Purity 98%
Chemical Name or Material Quinoline-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.