accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)NH2(S)H3CO

CAS RN 6485-55-8

IUPAC Name: (2R,6S)-2,6-dimethylmorpholin-4-ium
Synonyms: (2R,6S)-2,6-dimethylmorpholin-4-ium
Molecular Weight (g/mol): 116.18
Molecular Formula: C6H14NO
InChi Key: HNVIQLPOGUDBSU-OLQVQODUSA-O
SMILES: C[C@H]1C[NH2+]C[C@@H](C)O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results