missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine, TRC

Product Code. 30373645
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373645 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373645 Supplier Toronto Research Chemicals Supplier No. TRCD47474550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material cis-2,6-Dimethylmorpholine
CAS 6485-55-8
Molecular Formula C6H13NO
Quantity 50 g
Synonym (2R,6S)-rel-2,6-Dimethylmorpholine, (2R,6S)-2,6-Dimethylmorpholine, (2R,6S)-2,6-Dimethylmorpholine, 2,6-cis-Dimethylmorpholine, cis-(2R,6S)-2,6-Dimethylmorpholine, cis-2,6-Dimethylmorpholine
InChI Formula InChI=1S/C6H13NO/c1-5-3-7-4-6(2)8-5/h5-7H,3-4H2,1-2H3/t5-,6+
SMILES C[C@@H]1CNC[C@H](C)O1
IUPAC Name (2R,6S)-2,6-dimethylmorpholine
Molecular Weight (g/mol) 115.07
Formula Weight 115.0997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.