accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSOO

CAS RN 65085-83-8

IUPAC Name: 1-[4-(benzenesulfonyl)phenyl]ethan-1-one
Synonyms: 1-[4-(benzenesulfonyl)phenyl]ethanone
Molecular Weight (g/mol): 260.31
Molecular Formula: C14H12O3S
InChi Key: FGFHDNIGKVTTLC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results