accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 651-97-8

FOHOOHOF

CAS RN 651-97-8

IUPAC Name: 3,6-difluorobenzene-1,2-dicarboxylic acid
Synonyms: 3,6-difluorobenzene-1,2-dicarboxylic acid
Molecular Weight (g/mol): 202.11
Molecular Formula: C8H4F2O4
InChi Key: VFLMWMTWWZGXGA-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(F)=C1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results