Learn More
3,6-Difluorophthalic Acid, TRC
Product Code. 30369680
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369680 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30369680 Supplier Toronto Research Chemicals Supplier No. TRCD4481202.5G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,6-Difluorophthalic Acid
CAS 651-97-8
Molecular Formula C8H4F2O4
Quantity 2.5 g
Synonym 3,6-Difluoro-1,2-benzenedicarboxylic Acid
InChI Formula InChI=1S/C8H4F2O4/c9-3-1-2-4(10)6(8(13)14)5(3)7(11)12/h1-2H,(H,11,12)(H,13,14)
SMILES OC(=O)c1c(F)ccc(F)c1C(=O)O
IUPAC Name 3,6-difluorophthalic acid
Molecular Weight (g/mol) 202.11
Formula Weight 202.0078