accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBrHOBr(?)

CAS RN 65355-08-0

IUPAC Name: 3,3'-dibromo-5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Synonyms: 3,3'-dibromo-5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Molecular Weight (g/mol): 452.19
Molecular Formula: C20H20Br2O2
InChi Key: WDRTXCNGVVLRSZ-UHFFFAOYSA-N
SMILES: OC1=C(C2=C(CCCC2)C=C1Br)C1=C2CCCCC2=CC(Br)=C1O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)