missing translation for 'onlineSavingsMsg'
Learn More

(R)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol, 97%, BLD Pharm™

Product Code. 31433305
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31433305 25 g
31418168 100 mg
31421851 5 g
31419424 250 mg
31417334 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31433305 Supplier BLD Pharm Supplier No. BD119725g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 65355-08-0
Molecular Formula C20H20Br2O2
MDL Number MFCD03093977
Quantity 25 g
SMILES OC1=C(Br)C=C2CCCCC2=C1C3=C4CCCCC4=CC(Br)=C3O
Molecular Weight (g/mol) 452.18
Percent Purity 97%
Chemical Name or Material (R)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.