accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 65873-72-5

IUPAC Name: 6-methoxypyridine-3-carbaldehyde
Synonyms: 6-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: CTAIEPPAOULMFY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=O)C=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)