accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO

CAS RN 66-25-1

IUPAC Name: hexanal
Synonyms: hexanal n-caproic aldehyde fema number 2557 n-capronaldehyde hexaldehyde (substance) caproaldehyde caproic aldehyde n-caproaldehyde aldehyde C-6 n-caproylaldehyde
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: JARKCYVAAOWBJS-UHFFFAOYSA-N
SMILES: CCCCCC=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)