accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66-81-9

IUPAC Name: 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Synonyms: cycloheximide cicloheximida cycloheximid zykloheximid cicloheximidum
Molecular Weight (g/mol): 281.35
Molecular Formula: C15H23NO4
InChi Key: YPHMISFOHDHNIV-FSZOTQKASA-N
SMILES: C[C@H]1C[C@H](C)C(=O)[C@@H](C1)[C@H](O)CC1CC(=O)NC(=O)C1
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (4)
Color 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)