missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Cycloheximide ReadyMade™ Solution

Product Code. 30044037
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044037 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 30044037 Supplier TOKU-E™ Supplier No. C08410x1ML
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Cycloheximide ReadyMade™ Solution is provided as a solution of Cycloheximide in DMSO.

  • CAS Number: 66-81-9
  • Molecular Formula: C15H23NO4
  • Molecular Weight: 281.35 g/mol
  • Storage Conditions: 2-8°C
  • Form: Liquid
  • Source: Streptomyces griseus
  • Spectrum: Cycloheximide is effective against fungi and yeast, including fungi found in brewing test media. It has lower activity against bacteria
  • Mechanism of Action: Cycloheximide binds to the ribosome and inhibits the eEF2-mediated translocation step in protein synthesis, thus blocking translational elongation

Specifications

CAS 66-81-9
Certifications/Compliance Purity >95%
Chemical Name or Material Cycloheximide
Color Clear to Light yellow
Content And Storage 2°C to 8°C
Description Cycloheximide ReadyMade™ Solution 100 mg/mL in DMSO
Concentration 100 to 105 mg/mL
Form Clear, colorless to slight yellow solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid in DMSO
For Use With (Application) Microbiology, Plant Biology, Eukaryotic Cell Culture, Cancer Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >95%
Quantity 10 x 1 mL
Shipping Condition Ambient
Source Streptomyces griseus
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.