accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3OHNOO

CAS RN 66108-30-3

IUPAC Name: 1-(2-hydroxy-5-methyl-3-nitrophenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-5-methyl-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: XSHQMMIEZHWNAK-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(C)=CC(=C1O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results