CAS RN 666-52-4
Acetone-d6, for NMR, with 0.03% TMS, 99.9 atom% D
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, for NMR, 99+ atom % D
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, for NMR, 99.5 atom % D
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, for NMR, 99.8 atom % D, AcroSeal™
Acetone-d6, for NMR, packaged in 0.75ml ampoules, 100.0 atom % D
Acetone-d{6}, 99.9% (Isotopic)
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, for NMR, contains 1 v/v% TMS, 99.5 atom % D
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d{6}, 100%(Isotopic), contains 0.03% v/v TMS
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D
Acetone-d6, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Acetone-d6, for NMR, 99.8 atom % D
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d{6}, 99.9% (Isotopic), contains 1% v/v TMS
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, 100%(Isotopic), Thermo Scientific™
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
Acetone-d6, 100% (Isotopic), Thermo Scientific™
CAS: 666-52-4 Molecular Formula: C3H6O Molecular Weight (g/mol): 64.117 MDL Number: MFCD00044635 InChI Key: CSCPPACGZOOCGX-WFGJKAKNSA-N Synonym: acetone-d6,2h6 acetone,hexadeuteroacetone,acetone d6,perdeuteroacetone,deuterated acetone,unii-b0n19b53h8,cd3 2co,2-propanone-1,1,1,3,3,3-d6,d6-acetone PubChem CID: 522220 ChEBI: CHEBI:78217 IUPAC Name: 1,1,1,3,3,3-hexadeuteriopropan-2-one SMILES: CC(=O)C
- 1
- 2