accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNNNCH3

CAS RN 669707-13-5

IUPAC Name: 5,6-diethyl-3-(pyridin-2-yl)-1,2,4-triazine
Synonyms: 5,6-diethyl-3-(pyridin-2-yl)-1,2,4-triazine
Molecular Weight (g/mol): 214.27
Molecular Formula: C12H14N4
InChi Key: CTHOVPODUCLZPG-UHFFFAOYSA-N
SMILES: CCC1=C(CC)N=C(N=N1)C1=CC=CC=N1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results