accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 66992-27-6

IUPAC Name: sodium 2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonate
Synonyms: sodium 2-[bis(2-hydroxyethyl)amino]ethanesulfonate
Molecular Weight (g/mol): 235.23
Molecular Formula: C6H14NNaO5S
InChi Key: CFQLQLSIZOWFNV-UHFFFAOYSA-M
SMILES: [Na+].OCCN(CCO)CCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (11)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (6)
Concentration 
  • (4)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (8)
  • (3)
  • (11)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (6)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)