missing translation for 'onlineSavingsMsg'
Learn More

BES-buffered saline (2X), Thermo Scientific™

Product Code. 15494429
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15494429

Brand: Thermo Scientific Alfa Aesar J61955.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66992-27-6
Molecular Formula C6H14NNaO5S
Molecular Weight (g/mol) 235.23
MDL Number MFCD00217173
InChI Key CFQLQLSIZOWFNV-UHFFFAOYSA-M
PubChem CID 23671640
IUPAC Name sodium;2-[bis(2-hydroxyethyl)amino]ethanesulfonate
SMILES C(CO)N(CCO)CCS(=O)(=O)[O-].[Na+]

Specifications

Concentration 2X
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material BES-buffered saline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.