accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O(E)

CAS RN 6728-31-0

IUPAC Name: (4E)-hept-4-enal
Synonyms: cis-4-heptenal
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: VVGOCOMZRGWHPI-ONEGZZNKSA-N
SMILES: CC\C=C\CCC=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results