accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClFONNF

CAS RN 67469-78-7

IUPAC Name: dihydrogen 1-{2-[bis(4-fluorophenyl)methoxy]ethyl}-4-(3-phenylpropyl)piperazine dichloride
Synonyms: vanoxerine
Molecular Weight (g/mol): 523.49
Molecular Formula: C28H34Cl2F2N2O
InChi Key: MIBSKSYCRFWIRU-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].FC1=CC=C(C=C1)C(OCCN1CCN(CCCC2=CC=CC=C2)CC1)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results