accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCH3

CAS RN 67589-52-0

IUPAC Name: 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate
Synonyms: 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate
Molecular Weight (g/mol): 304.43
Molecular Formula: C19H28O3
InChi Key: HJIVFZUPTYADSP-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C(=O)OC1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results