missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 97%, Thermo Scientific™

Product Code. 15462296
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462296 5 g 5g
15452296 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462296 Supplier Thermo Scientific Alfa Aesar Supplier No. H55792.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (1.7E-3 g/L) (25°C).

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67589-52-0
Molecular Formula C19H28O3
Molecular Weight (g/mol) 304.43
MDL Number MFCD11053432
InChI Key HJIVFZUPTYADSP-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, 4-pentyl-, 4-methoxyphenyl ester, trans, trans-4-methoxyphenyl 4-pentylcyclohexanecarboxylate, trans-4-methoxy-phenyl 4-pentyl-cyclohexanecarboxylate, trans-4-pentylcyclohexanecarboxylic acid, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-pentylcyclohexanoate, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate, 4-methoxyphenyl trans-4-pentylcyclohexanecarboxylate, hjivfzuptyadsp-wkilwmfisa-n, 4-methoxyphenyl 4-pentylcyclohexanecarboxylate #, 4-methoxyphenyl 4-pentylcyclohexane-1-carboxylate
PubChem CID 105460
IUPAC Name (4-methoxyphenyl) 4-pentylcyclohexane-1-carboxylate
SMILES CCCCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC

Specifications

Melting Point 40°C to 44°C
Quantity 5 g
Solubility Information Sparingly Soluble in water (1.7E-3g/L) (25°C).
Formula Weight 304.43
Percent Purity 97%
Chemical Name or Material 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.