accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OFFOH

CAS RN 676500-39-3

IUPAC Name: 2,3-difluoro-4-hydroxybenzaldehyde
Synonyms: 2,3-difluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: KTGPDDPEVRJSFY-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(C=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results