accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOF

CAS RN 677728-92-6

IUPAC Name: 6-fluoropyridine-3-carbaldehyde
Synonyms: 6-fluoropyridine-3-carbaldehyde
Molecular Weight (g/mol): 125.1
Molecular Formula: C6H4FNO
InChi Key: PZPNGWWKCSJKOS-UHFFFAOYSA-N
SMILES: FC1=NC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results