accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 67808-66-6

CH3OOSO

CAS RN 67808-66-6

IUPAC Name: methyl 5-formylthiophene-3-carboxylate
Synonyms: methyl 5-formylthiophene-3-carboxylate
Molecular Weight (g/mol): 170.18
Molecular Formula: C7H6O3S
InChi Key: GNXNZRYWBFMVHK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CSC(C=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results