accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6834-92-0

NaNaOSiOO

CAS RN 6834-92-0

IUPAC Name: disodium oxosilanebis(olate)
Synonyms: sodium silicate sodium polysilicate sodium sesquisilicate sodium siliconate sodium metasilicate sodium silicate glass
Molecular Weight (g/mol): 122.06
Molecular Formula: Na2O3Si
InChi Key: NTHWMYGWWRZVTN-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][Si]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)