missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metasilicate, TRC

Product Code. 30412519
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412519 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412519 Supplier Toronto Research Chemicals Supplier No. TRCS22440510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Metasilicate
CAS 6834-92-0
Molecular Formula Na2O3Si
Quantity 10 g
Synonym Blue Meta, Crystamet, Disodium Metasilicate, Disodium Monosilicate, Disodium Silicate, Drymet 59, Mead Beads 2048, Metso 20, Metso 2048, Metso 510, Metso Pentabead 20, Metso-Beads 2048, Orthosil, P 84, P 84 (silicate), Purifeed 6N, R-SIL, S 108358, SMS, SP 20, SP 20 (silicate), Silicon Sodium Oxide (SiNa2O3), Simet 5G, Simet A, Simet AG, Simet AP, Simet GA 5, Sodium Metasilicate Anhydrous, Granular D, Sodium Metasilicate, Sodium Metasilicate (Na2SiO3), Sodium Silicate (Na2(SiO3)), Sodium Silicate (Na2SiO3), Sodium Silicate (Na6(SiO3)3), Sodium Silicon Oxide (Na2SiO3), Starso, Stixso NN
InChI Formula InChI=1S/2Na.O3Si/c;;1-4(2)3/q2*+1;-2
SMILES [Na+].[Na+].[O-][Si](=O)[O-]
IUPAC Name disodium;dioxido(oxo)silane
Molecular Weight (g/mol) 122.06
Formula Weight 121.9412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.