accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6846-50-0

IUPAC Name: 2,2,4-trimethyl-1-[(2-methylpropanoyl)oxy]pentan-3-yl 2-methylpropanoate
Synonyms: trimethyl pentanyl diisobutyrate
Molecular Weight (g/mol): 286.41
Molecular Formula: C16H30O4
InChi Key: OMVSWZDEEGIJJI-UHFFFAOYNA-N
SMILES: CC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)