missing translation for 'onlineSavingsMsg'
Learn More

2,2,4-Trimethyl-1,3-pentanediol Diisobutyrate, TRC

Product Code. 30453761
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453761 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453761 Supplier Toronto Research Chemicals Supplier No. TRCT79812025G
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate (TXIB)
CAS 6846-50-0
Molecular Formula C16 H30 O4
Quantity 25 g
Synonym Propanoic acid','2-methyl-','1,1'-[2,2-dimethyl-1-(1-methylethyl)-1,3-propanediyl] ester (ACI), 1,3-Pentanediol','2,2,4-trimethyl-','diisobutyrate (8CI), Isobutyric acid','1-isopropyl-2,2-dimethyltrimethylene ester (7CI','8CI), Propanoic acid','2-methyl-','2,2-dimethyl-1-(1-methylethyl)-1,3-propanediyl ester (9CI), 1-Isopropyl-2,2-dimethyltrimethylene diisobutyrate, 2,2,4-Trimethyl-1,3-pentanediol 2-methylpropanoic acid diester, 2,2,4-Trimethyl-1,3-pentanediol bis(isobutyrate), 2,2,4-Trimethyl-1,3-pentanediol di(isopropylformate), 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 2,2,4-Trimethyl-1,3-pentanediyl diisobutyrate, 2,2,4-Trimethylpentane 1,3-diisobutanoate, 2,2-Dimethyl-1-(1-methylethyl)-1,3-propanediyl bis(2-methylpropanoate), [2,2,4-Trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate, C 16, CS 16, Eastman TXIB, Kodaflex TXIB, Kyowanol D, Kyowanol M, NX 800, OE 300, Optifilm Enhancer 300, RTC 16, Texanol isobutyrate, Texanol OE 300, TXIB
InChI Formula InChI=1S/C16H30O4/c1-10(2)13(20-15(18)12(5)6)16(7,8)9-19-14(17)11(3)4/h10-13H,9H2,1-8H3
SMILES CC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C
IUPAC Name [2,2,4-trimethyl-3-(2-methylpropanoyloxy)pentyl] 2-methylpropanoate
Molecular Weight (g/mol) 286.41
Formula Weight 286.2144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.