accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOONCOCl

CAS RN 68622-16-2

IUPAC Name: 1-chloro-2-isocyanato-4-nitrobenzene
Synonyms: 1-chloro-2-isocyanato-4-nitrobenzene
Molecular Weight (g/mol): 198.56
Molecular Formula: C7H3ClN2O3
InChi Key: DZMSHKRHIIDOIZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(N=C=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results