CAS RN 6892-68-8
Dithioerythritol, 99+%
CAS: 6892-68-8 Molecular Formula: C4H10O2S2 Molecular Weight (g/mol): 154.24 MDL Number: MFCD00063750 InChI Key: VHJLVAABSRFDPM-ZXZARUISSA-N Synonym: dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol PubChem CID: 439352 ChEBI: CHEBI:17456 SMILES: O[C@@H](CS)[C@H](O)CS
1,4-Dithioerythritol, Electrophoresis Grade, 99%
CAS: 6892-68-8 Molecular Formula: C4H10O2S2 Molecular Weight (g/mol): 154.24 MDL Number: MFCD00063750 InChI Key: VHJLVAABSRFDPM-ZXZARUISSA-N Synonym: dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol PubChem CID: 439352 ChEBI: CHEBI:17456 SMILES: O[C@@H](CS)[C@H](O)CS
1,4-Dithioerythritol, 98+%
CAS: 6892-68-8 Molecular Formula: C4H10O2S2 Molecular Weight (g/mol): 154.24 MDL Number: MFCD00063750 InChI Key: VHJLVAABSRFDPM-ZXZARUISSA-N Synonym: dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol PubChem CID: 439352 ChEBI: CHEBI:17456 SMILES: O[C@@H](CS)[C@H](O)CS
Dithioerythritol, MP Biomedicals™
CAS: 6892-68-8 Molecular Formula: C4H10O2S2 Molecular Weight (g/mol): 154.24 MDL Number: MFCD00063750 InChI Key: VHJLVAABSRFDPM-ZXZARUISSA-N Synonym: dithioerythritol,1,4-dithioerythritol,dte,2r,3s-1,4-dimercaptobutane-2,3-diol,erythro-1,4-dimercapto-2,3-butanediol,2,3-butanediol, 1,4-dimercapto-, 2r,3s-rel,1,4-dithioerythritol dte,2r*,3s*-1,4-dimercapto-2,3-butanediol,erythro-2,3-dihydroxy-1,4-butanedithiol,2r,3s-1,4-disulfanylbutane-2,3-diol PubChem CID: 439352 ChEBI: CHEBI:17456 SMILES: O[C@@H](CS)[C@H](O)CS
1,4-Dithioerythritol, TRC
CAS: 6892-68-8 Molecular Formula: C4 H10 O2 S2 Molecular Weight (g/mol): 154.25 Synonym: 1,4-Dithioerythritol,erythro-1,4-Dimercapto-2,3-butanediol,Cleland's reagent,Dithioerythritol,DTE IUPAC Name: 1,4-bis(sulfanyl)butane-2,3-diol SMILES: OC(CS)C(O)CS
Melford DTE
Protective reagent for maintaining -SH groups in reduced stated.