accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 69056-38-8

IUPAC Name: dihydrogen (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-1,4,5,6,7,8-hexahydropteridin-4-one dichloride
Synonyms: dihydrogen tetrahydrobiopterin dichloride
Molecular Weight (g/mol): 314.17
Molecular Formula: C9H17Cl2N5O3
InChi Key: RKSUYBCOVNCALL-NTVURLEBSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].C[C@H](O)[C@H](O)[C@H]1CNC2=C(N1)C(=O)N=C(N)N2
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results