missing translation for 'onlineSavingsMsg'
Learn More

(6R)-5,6,7,8-Tetrahydrobiopterin Dihydrochloride, TRC

Product Code. 30180324
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180324 25 mg 25mg
30179897 5 mg 5mg
30179824 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180324 Supplier Toronto Research Chemicals Supplier No. TRCT79561325MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H15N5O3 . 2(HCl)
Molecular Weight (g/mol) 314.17
Synonym BioMarin T 1401, Biopten, Kuvan, SUN 0588, Sapropterin Dihydrochloride, Shiratori SN 0588
IUPAC Name (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
SMILES Cl.Cl.C[C@H](O)[C@H](O)[C@H]1CNC2=C(N1)C(=O)N=C(N)N2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C9H15N5O3.2ClH/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7;;/h3-4,6,12,15-16H,2H2,1H3,(H4,10,11,13,14,17);2*1H/t3-,4+,6-;;/m0../s1
Formula Weight 313.0708
Percent Purity >95%
Chemical Name or Material (6R)-5,6,7,8-Tetrahydrobiopterin Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.